Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161763.1 | NP_001155235.1 | 3 | 426 | ||
NM_001161765.1 | NP_001155237.1 | 2 | 509 | |||
NM_010232.4 | NP_034362.2 | 3 | 410 | |||
GenBank mRNA | AK133675.1 | - | 2 | 491 | ||
AK147040.1 | - | 3 | 402 | |||
AK162951.1 | - | 3 | 427 | |||
BC022991.1 | - | 3 | 373 | 5123876 | ||
CN664821.1 | - | 3 | 410 |