Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010243.3 | NP_034373.1 | 3 | 768 | ||
GenBank mRNA | AB015426.1 | - | 3 | 372 | ||
AK032177.1 | - | 3 | 910 | |||
AK033607.1 | - | 2 | 923 | |||
AK036183.1 | - | 3 | 571 | |||
AK043410.1 | - | 3 | 609 | |||
AK047650.1 | - | 3 | 567 | |||
AK049122.1 | - | 4 | 849 | |||
AK163871.1 | - | 3 | 568 | |||
BC116874.1 | - | 3 | 464 | 40126193 | ||
BC116876.1 | - | 3 | 464 | 40126195 |