Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010260.1 | NP_034390.1 | 5 | 619 | ||
GenBank mRNA | AF077007.1 | - | 5 | 619 | ||
AF109168.1 | - | 5 | 572 | |||
AJ007970.1 | - | 5 | 612 | |||
AK150138.1 | - | 5 | 555 | |||
BC011336.1 | - | 5 | 608 | 3494311 | ||
BC032882.1 | - | 5 | 634 | 1230883 | ||
BC049141.1 | - | 5 | 597 | |||
CT010249.1 | - | 4 | 449 |