Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010269.3 | NP_034399.1 | 3 | 508 | ||
GenBank mRNA | AK004887.1 | - | 3 | 454 | ||
AK082958.1 | - | 3 | 470 | |||
AK146835.1 | - | 3 | 498 | |||
AK161122.1 | - | 3 | 445 | |||
AK166799.1 | - | 3 | 423 | |||
AK166859.1 | - | 3 | 422 | |||
BC025070.1 | - | 3 | 417 | 3710712 | ||
Y17851.1 | - | 3 | 506 |