Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301332.1 | NP_001288261.1 | 3 | 2994 | ||
NM_001301333.1 | NP_001288262.1 | 3 | 2356 | |||
NM_001301357.1 | NP_001288286.1 | 2 | 2044 | |||
NM_010275.3 | NP_034405.1 | 3 | 3072 | |||
GenBank mRNA | U37459.1 | - | 3 | 2109 |