Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285457.1 | NP_001272386.1 | 9 | 1729 | ||
NM_010279.3 | NP_034409.1 | 11 | 1945 | |||
GenBank mRNA | AK076171.1 | - | 11 | 1866 | ||
AK139987.1 | - | 6 | 786 | |||
BC011532.1 | - | 9 | 1676 | |||
BC040251.1 | - | 9 | 1513 | |||
BC044783.2 | - | 12 | 1911 | |||
BC054378.1 | - | 10 | 1616 | 6390018 | ||
KM079499.1 | - | 10 | 1604 |