Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP4R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042388.2 | NP_001035847.1 | 6 | 574 | ||
NM_005134.3 | NP_005125.1 | 6 | 523 | |||
NR_052003.1 | 5 | 416 | ||||
XM_011525775.2 | XP_011524077.1 | 6 | 700 | |||
XM_011525776.1 | XP_011524078.1 | 6 | 564 | |||
XM_011525779.2 | XP_011524081.1 | 6 | 700 | |||
XM_017026105.1 | XP_016881594.1 | 6 | 700 | |||
XM_017026106.1 | XP_016881595.1 | 5 | 378 | |||
XM_017026107.1 | XP_016881596.1 | 5 | 393 | |||
GenBank mRNA | AF111106.1 | - | 6 | 493 | ||
AF200478.1 | - | 6 | 473 | |||
AK027064.1 | - | 6 | 525 | |||
AK292538.1 | - | 6 | 454 | |||
AK293869.1 | - | 5 | 314 | |||
AK315106.1 | - | 6 | 481 | |||
BC060829.1 | - | 6 | 475 | 30342594 | ||
DR006046.1 | - | 6 | 530 |