Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gykl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010293.3 | NP_034423.2 | 1 | 1147 | ||
GenBank mRNA | AF117733.1 | - | 1 | 1147 | ||
AK076631.1 | - | 1 | 1123 | |||
BC050968.1 | - | 1 | 1136 | |||
BC119574.2 | - | 1 | 1128 | 40058020 |