Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281969.1 | NP_001268898.1 | 6, 7 | 739 | ||
NM_010298.6 | NP_034428.2 | 6, 7 | 789 | |||
GenBank mRNA | AK082578.1 | - | 6, 7 | 792 | ||
AK083251.1 | - | 5, 6 | 643 | |||
AM420316.1 | - | 5, 6 | 622 | |||
BC027094.1 | - | 6, 7 | 743 | |||
BC037605.1 | - | 6, 7 | 757 | 5361798 | ||
U09399.1 | - | 6, 7 | 698 | |||
X81202.1 | - | 5, 6 | 581 |