Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010299.3 | NP_034429.1 | 2 | 336 | ||
GenBank mRNA | AK149642.1 | - | 2 | 228 | ||
AK152256.1 | - | 2 | 228 | |||
AK179394.1 | - | 2 | 144 | |||
AK181597.1 | - | 2 | 144 | |||
BC004651.1 | - | 2 | 204 | 3482848 | ||
CT010289.1 | - | 2 | 157 | |||
L19526.1 | - | 2 | 170 |