Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gna13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010303.3 | NP_034433.3 | 4 | 973 | ||
GenBank mRNA | AK149766.1 | - | 4 | 867 | ||
BC057665.1 | - | 4 | 827 | 4918717 | ||
M57620.1 | - | 1 | 112 | |||
M63660.1 | - | 4 | 921 |