Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010306.3 | NP_034436.1 | 3 | 423 | ||
GenBank mRNA | AK004566.1 | - | 3 | 417 | ||
AK077490.1 | - | 3 | 385 | |||
AK133749.1 | - | 3 | 364 | |||
AK167438.1 | - | 3 | 377 | |||
AK168578.1 | - | 3 | 361 | |||
AK169067.1 | - | 3 | 368 | |||
AK169285.1 | - | 3 | 398 | |||
AK187104.1 | - | 3 | 188 | |||
BC003462.1 | - | 3 | 351 | |||
BC020009.1 | - | 3 | 395 | |||
BC041107.1 | - | 3 | 334 | 2648164 | ||
S79610.1 | - | 2 | 148 |