Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnpat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010322.3 | NP_034452.3 | 16 | 2893 | ||
GenBank mRNA | AK150810.1 | - | 16 | 2851 | ||
AK166997.1 | - | 16 | 2865 | |||
BC025972.1 | - | 16 | 2850 | 4020505 | ||
BY469630.1 | - | 1 | 373 |