Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpar1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290486.1 | NP_001277415.1 | 2 | 541 | ||
NM_010336.2 | NP_034466.2 | 3 | 656 | |||
NM_172989.1 | NP_766577.1 | 3 | 745 | |||
GenBank mRNA | AK030330.1 | - | 3 | 709 | ||
BC025425.1 | - | 3 | 655 | 5009055 | ||
U48235.1 | - | 2 | 506 | |||
U70622.1 | - | 3 | 697 |
Set Membership: |
G-protein coupled receptor |