Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135110.2 | NP_001128582.1 | 3 | 463 | ||
NM_005147.5 | NP_005138.3 | 3 | 463 | |||
GenBank mRNA | AB209923.1 | - | 3 | 382 | ||
AF061749.1 | - | 3 | 417 | |||
AF244136.1 | - | 3 | 388 | |||
AF411044.1 | - | 3 | 430 | |||
AK001333.1 | - | 3 | 403 | |||
AK223105.1 | - | 3 | 403 | |||
AK314218.1 | - | 3 | 400 | |||
BC007225.1 | - | 3 | 383 | |||
BC011855.2 | - | 3 | 387 | 4139881 | ||
BC012343.1 | - | 3 | 379 | 4653507 | ||
BC014062.2 | - | 3 | 379 | |||
BC030145.2 | - | 3 | 379 | |||
BC032100.2 | - | 3 | 401 | 4660602 | ||
BX445320.2 | - | 3 | 463 | |||
DA172592.1 | - | 3 | 411 |