Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010343.2 | NP_034473.2 | 5 | 1119 | ||
GenBank mRNA | AK033841.1 | - | 2 | 713 | ||
BC100749.1 | - | 5 | 1113 | 40046457 | ||
BC103588.1 | - | 5 | 1113 | 40045838 | ||
BC103589.1 | - | 5 | 1115 | 40045837 | ||
BC105648.1 | - | 5 | 1114 | 40046458 | ||
X53780.1 | - | 4 | 951 |