Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177629.1 | NP_001171100.1 | 4 | 434 | ||
NM_010345.4 | NP_034475.2 | 4 | 594 | |||
GenBank mRNA | AF022072.1 | - | 4 | 568 | ||
AK012646.1 | - | 4 | 574 | |||
AK030727.1 | - | 4 | 529 | |||
AK032927.1 | - | 4 | 575 | |||
AK163841.1 | - | 4 | 575 | |||
BC016111.1 | - | 4 | 434 | 4481345 | ||
BC053842.1 | - | 4 | 561 |