Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177629.1 | NP_001171100.1 | 9 | 1082 | ||
NM_010345.4 | NP_034475.2 | 10 | 1407 | |||
GenBank mRNA | AF022072.1 | - | 10 | 1381 | ||
AK030727.1 | - | 10 | 1342 | |||
AK032927.1 | - | 10 | 1388 | |||
AK163841.1 | - | 10 | 1388 | |||
AK169000.1 | - | 4 | 371 | |||
BC016111.1 | - | 9 | 1082 | 4481345 | ||
BC053842.1 | - | 10 | 1374 |