Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010346.2 | NP_034476.1 | 15 | 2165 | ||
GenBank mRNA | AK057640.1 | - | 15 | 2158 | ||
AK145996.1 | - | 15 | 1981 | |||
AK168438.1 | - | 15 | 1949 | |||
AK220277.1 | - | 14 | 2057 | |||
BC003295.1 | - | 15 | 1910 | 3583896 | ||
BC081547.1 | - | 14 | 2156 | |||
M94450.1 | - | 15 | 2186 |