Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gsg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080552.1 | NP_001074021.1 | 6 | 1056 | ||
NM_001080553.1 | NP_001074022.1 | 6 | 1209 | |||
NM_010352.2 | NP_034482.2 | 7 | 1269 | |||
GenBank mRNA | AK006326.1 | - | 4 | 652 | ||
AK088285.1 | - | 4 | 652 | |||
BC023009.1 | - | 6 | 1141 | 5364798 | ||
D87325.1 | - | 7 | 1144 |