Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005159.4 | NP_005150.1 | 3 | 407 | ||
GenBank mRNA | AK056592.1 | - | 3 | 230 | ||
AK129892.1 | - | 3 | 230 | |||
AK129902.1 | - | 3 | 230 | |||
AK290616.1 | - | 3 | 230 | |||
BC009978.2 | - | 3 | 369 | 4121697 | ||
CR541795.1 | - | 2 | 164 |