Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077696.1 | NP_001071164.1 | 8 | 1133 | ||
NM_001284248.1 | NP_001271177.1 | 10 | 1645 | |||
NM_001284249.1 | NP_001271178.1 | 9 | 1242 | |||
NM_001284250.1 | NP_001271179.1 | 8 | 1130 | |||
NM_010412.3 | NP_034542.3 | 8 | 1130 | |||
GenBank mRNA | AF006602.2 | - | 7 | 846 | ||
AF207748.1 | - | 7 | 765 | |||
AK122325.2 | - | 3 | 169 | |||
AK139202.1 | - | 9 | 1216 | |||
AK145972.1 | - | 8 | 1096 | |||
AK146483.1 | - | 8 | 1069 | |||
AK155335.1 | - | 8 | 1104 | |||
AK156008.1 | - | 5 | 1381 | |||
AK157159.1 | - | 5 | 1381 | |||
BC049926.1 | - | 8 | 1066 | |||
BC058554.1 | - | 10 | 1597 | |||
BC060609.1 | - | 7 | 924 | 5720438 |