Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077696.1 | NP_001071164.1 | 14, 15 | 2405 | ||
NM_001284248.1 | NP_001271177.1 | 16, 17 | 2917 | |||
NM_001284249.1 | NP_001271178.1 | 15, 16 | 2514 | |||
NM_001284250.1 | NP_001271179.1 | 14, 15 | 2402 | |||
NM_010412.3 | NP_034542.3 | 14, 15 | 2402 | |||
GenBank mRNA | AF006602.2 | - | 13, 14 | 2115 | ||
AF207748.1 | - | 13, 14 | 2034 | |||
AK122325.2 | - | 9, 10 | 1444 | |||
AK139202.1 | - | 15, 16 | 2488 | |||
AK145972.1 | - | 14, 15 | 2368 | |||
AK146483.1 | - | 14, 15 | 2341 | |||
AK155335.1 | - | 14, 15 | 2376 | |||
AK156008.1 | - | 8, 9 | 3239 | |||
AK157159.1 | - | 8, 9 | 3239 | |||
BC023309.1 | - | 5, 6 | 955 | |||
BC049926.1 | - | 14, 15 | 2336 | |||
BC058554.1 | - | 16, 17 | 2869 | |||
BC060609.1 | - | 13, 14 | 2196 | 5720438 |