Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077696.1 | NP_001071164.1 | 21 | 2993 | ||
NM_001284248.1 | NP_001271177.1 | 23 | 3505 | |||
NM_001284249.1 | NP_001271178.1 | 22 | 3102 | |||
NM_001284250.1 | NP_001271179.1 | 21 | 2990 | |||
NM_010412.3 | NP_034542.3 | 21 | 2990 | |||
GenBank mRNA | AF006602.2 | - | 20 | 2703 | ||
AF207748.1 | - | 20 | 2622 | |||
AK122325.2 | - | 16 | 2032 | |||
AK139202.1 | - | 22 | 3076 | |||
AK145972.1 | - | 21 | 2956 | |||
AK146483.1 | - | 21 | 2929 | |||
AK155335.1 | - | 21 | 2964 | |||
AK156008.1 | - | 15 | 3827 | |||
AK157159.1 | - | 15 | 3827 | |||
AK161956.1 | - | 1 | 122 | |||
BC023309.1 | - | 12 | 1543 | |||
BC049926.1 | - | 21 | 2924 | |||
BC058554.1 | - | 23 | 3457 | |||
BC060609.1 | - | 20 | 2784 | 5720438 |