Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbegf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010415.2 | NP_034545.1 | 3 | 629 | ||
GenBank mRNA | AK145548.1 | - | 3 | 598 | ||
AK160575.1 | - | 3 | 599 | |||
AK164309.1 | - | 3 | 630 | |||
BC089607.1 | - | 3 | 598 | 6751441 | ||
L07264.1 | - | 3 | 562 |