Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hbegf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010415.2 | NP_034545.1 | 6 | 1207 | ||
GenBank mRNA | AK145548.1 | - | 6 | 1176 | ||
AK160575.1 | - | 6 | 1175 | |||
BC089607.1 | - | 6 | 1176 | 6751441 |