Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010418.2 | NP_034548.2 | 8, 9 | 1081 | ||
GenBank mRNA | AF061529.1 | - | 8, 9 | 1031 | ||
AF071173.1 | - | 8, 9 | 983 | |||
AK165121.1 | - | 7, 8 | 1024 | |||
AK216874.1 | - | 2, 3 | 257 |