Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hesx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010420.2 | NP_034550.2 | 1 | 440 | ||
GenBank mRNA | AK082831.1 | - | 1 | 88 | ||
BC132051.1 | - | 1 | 266 | 40130328 | ||
BC132053.1 | - | 1 | 266 | 40130330 | ||
BC145262.1 | - | 1 | 265 | 9053815 | ||
U40720.1 | - | 1 | 438 | |||
X80040.1 | - | 1 | 148 |