Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hexa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010421.4 | NP_034551.2 | 1, 2 | 310 | ||
GenBank mRNA | AK075895.1 | - | 1, 2 | 286 | ||
AK075911.1 | - | 1, 2 | 311 | |||
AK080777.1 | - | 1, 2 | 283 | |||
AK144168.1 | - | 1, 2 | 438 | |||
AK150444.1 | - | 1, 2 | 281 | |||
AK151960.1 | - | 1, 2 | 285 | |||
AK159091.1 | - | 1, 2 | 302 | |||
AK159814.1 | - | 1, 2 | 296 | |||
AK159890.1 | - | 1, 2 | 283 | |||
AK160112.1 | - | 1, 2 | 270 | |||
AK167013.1 | - | 1, 2 | 296 | |||
AK168186.1 | - | 1, 2 | 310 | |||
AK170903.1 | - | 1, 2 | 296 | |||
BC010755.1 | - | 1, 2 | 279 | 4225233 | ||
U07631.1 | - | 1, 2 | 302 | |||
X64331.1 | - | 1, 2 | 253 |