Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hira | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010435.2 | NP_034565.2 | 6 | 1190 | ||
GenBank mRNA | BC052856.1 | - | 6 | 608 | ||
BC060181.1 | - | 6 | 874 | |||
X75295.1 | - | 7 | 705 | |||
X92590.1 | - | 6 | 677 | |||
X99712.1 | - | 6 | 517 |