Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hira | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010435.2 | NP_034565.2 | 7 | 1318 | ||
GenBank mRNA | BC052856.1 | - | 7 | 736 | ||
BC060181.1 | - | 7 | 1002 | |||
X75295.1 | - | 8 | 833 | |||
X92590.1 | - | 7 | 805 | |||
X99712.1 | - | 7 | 645 |