Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005185.3 | NP_005176.1 | 1 | 613 | ||
GenBank mRNA | AK313934.1 | - | 1 | 541 | ||
BC031889.1 | - | 1 | 521 | 4777524 | ||
M36707.1 | - | 1 | 1481 | |||
M58026.1 | - | 1 | 541 |