Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAPN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198868.1 | NP_001185797.1 | 15 | 1828 | ||
NM_001198869.1 | NP_001185798.1 | 15 | 1797 | |||
NM_005186.3 | NP_005177.2 | 15 | 1813 | |||
NR_040008.1 | 14 | 1743 | ||||
XM_006718698.1 | XP_006718761.1 | 15 | 1728 | |||
XM_011545292.1 | XP_011543594.1 | 15 | 1739 | |||
GenBank mRNA | AK301538.1 | - | 14 | 1635 | ||
AK315556.1 | - | 15 | 1742 | |||
AY550975.1 | - | 14 | 1636 | |||
BC008751.2 | - | 15 | 1764 | 3347480 | ||
BC017200.2 | - | 15 | 1650 | 3629894 | ||
BC075862.1 | - | 15 | 1731 | 5223130 | ||
X04366.1 | - | 15 | 1774 |