Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005191.3 | NP_005182.1 | 3 | 523 | ||
XM_011513327.2 | XP_011511629.1 | 3 | 2589 | |||
XM_017007520.1 | XP_016863009.1 | 3 | 598 | |||
GenBank mRNA | AY081815.1 | - | 2 | 239 | ||
AY197777.1 | - | 2 | 128 | |||
AY197778.1 | - | 2 | 128 | |||
BC042665.1 | - | 3 | 491 | 5181343 | ||
BP225173.1 | - | 3 | 550 | |||
M27533.1 | - | 3 | 445 |