Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039129.4 | NP_001034218.1 | 11 | 1854 | ||
NM_010447.5 | NP_034577.1 | 10 | 1695 | |||
GenBank mRNA | AK088308.1 | - | 10 | 1638 | ||
AK131999.1 | - | 10 | 1662 | |||
AK135391.1 | - | 10 | 1640 | |||
AK146126.1 | - | 11 | 1795 | |||
AK146630.1 | - | 11 | 1820 | |||
AK152684.1 | - | 10 | 1637 | |||
AK161730.1 | - | 10 | 1639 | |||
AK166901.1 | - | 10 | 1639 | |||
AK166989.1 | - | 11 | 1796 | |||
AK167161.1 | - | 10 | 1639 | |||
AK167810.1 | - | 10 | 1637 | |||
AK167811.1 | - | 10 | 1636 | |||
AK167820.1 | - | 10 | 1641 | |||
AK167913.1 | - | 10 | 1637 | |||
AK168420.1 | - | 10 | 1640 | |||
AK168531.1 | - | 10 | 1642 | |||
AK168733.1 | - | 11 | 1798 | |||
AK169216.1 | - | 10 | 1640 | |||
BC052723.1 | - | 11 | 1782 | |||
BC080675.1 | - | 10 | 1621 | 6308746 | ||
BC089340.1 | - | 11 | 1774 | 6402927 | ||
BC092395.1 | - | 11 | 1777 | 6403376 | ||
D86728.1 | - | 9 | 1447 | |||
D86729.1 | - | 10 | 1603 |
Gene Symbol: Gm5643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BY428408.1 | - | 1 | 239 |