Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024652.4 | NP_078928.3 | 32 | 5995 | ||
XM_005254979.3 | XP_005255036.1 | 18 | 3653 | |||
XM_011522012.2 | XP_011520314.1 | 34 | 6269 | |||
XM_011522013.2 | XP_011520315.1 | 32 | 5978 | |||
XM_011522014.2 | XP_011520316.1 | 32 | 6193 | |||
XM_011522015.2 | XP_011520317.1 | 27 | 6788 | |||
XM_011522016.2 | XP_011520318.1 | 30 | 5470 | |||
XM_011522017.2 | XP_011520319.1 | 18 | 3660 | |||
XM_011522018.2 | XP_011520320.1 | 18 | 3655 | |||
XM_017022570.1 | XP_016878059.1 | 22 | 4242 | |||
GenBank mRNA | AB058693.1 | - | 18 | 3700 | ||
AK026772.1 | - | 1 | 370 | |||
AK054662.1 | - | 6 | 1443 | |||
AK058028.1 | - | 12 | 2543 | |||
AK094556.1 | - | 2 | 1123 | |||
AK131075.1 | - | 13 | 2928 | |||
AK225575.1 | - | 1 | 370 | |||
BC005408.1 | - | 7 | 1666 | |||
BC020379.1 | - | 7 | 1666 | |||
BC066655.1 | - | 3 | 908 | |||
BC110293.1 | - | 1 | 554 | |||
BC144705.1 | - | 30 | 5383 | |||
BC146769.1 | - | 18 | 3700 | |||
BC171802.1 | - | 31 | 5862 | |||
DQ013130.1 | - | 31 | 5633 |