Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010453.5 | NP_034583.1 | 1, 2 | 791 | ||
GenBank mRNA | AB221550.1 | - | 1, 2 | 554 | ||
AK144356.1 | - | 1, 2 | 532 | |||
BC011063.1 | - | 1, 2 | 615 | 3985274 | ||
M28021.1 | - | 1, 2 | 791 | |||
M36604.1 | - | 1, 2 | 668 | |||
X16840.1 | - | 1, 2 | 668 |