Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010461.2 | NP_034591.1 | 2 | 1701 | ||
GenBank mRNA | AB221615.1 | - | 2 | 640 | ||
AK131612.1 | - | 2 | 576 | |||
BC139209.1 | - | 2 | 881 | 8862231 | ||
BC139212.1 | - | 3 | 821 | 8862234 | ||
X13721.1 | - | 2 | 1700 |