Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010463.1 | NP_034593.1 | 1 | 284 | ||
GenBank mRNA | AB221553.1 | - | 1 | 284 | ||
AF448482.1 | - | 1 | 284 | |||
BC120845.1 | - | 1 | 380 | 40129768 | ||
BC120847.1 | - | 1 | 380 | 40129770 |