Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hp1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122896.2 | NP_001116368.1 | 2 | 198 | ||
NM_001122897.2 | NP_001116369.1 | 3 | 435 | |||
NM_001285478.1 | NP_001272407.1 | 3 | 455 | |||
NM_001285479.1 | NP_001272408.1 | 3 | 396 | |||
NM_001285480.1 | NP_001272409.1 | 2 | 198 | |||
NM_001285481.1 | NP_001272410.1 | 3 | 396 | |||
NM_010470.3 | NP_034600.2 | 3 | 396 | |||
GenBank mRNA | AK004938.1 | - | 3 | 373 | ||
AK028181.1 | - | 3 | 374 | |||
AK043260.1 | - | 2 | 172 | |||
AK050234.1 | - | 3 | 374 | |||
AK075725.1 | - | 2 | 176 | |||
AK150784.1 | - | 3 | 441 | |||
AK161041.1 | - | 2 | 166 | |||
AK161084.1 | - | 3 | 290 | |||
AK166161.1 | - | 3 | 382 | |||
AK169740.1 | - | 3 | 302 | |||
BC020024.1 | - | 3 | 346 | 3597151 | ||
BC107687.1 | - | 3 | 331 | |||
BU612149.1 | - | 2 | 238 | |||
BY130345.1 | - | 2 | 233 | |||
CN670198.1 | - | 2 | 237 | |||
X99642.1 | - | 1 | 39 |