Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hp1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122896.2 | NP_001116368.1 | 3 | 257 | ||
NM_001122897.2 | NP_001116369.1 | 4 | 494 | |||
NM_001285478.1 | NP_001272407.1 | 4 | 514 | |||
NM_001285479.1 | NP_001272408.1 | 4 | 455 | |||
NM_010470.3 | NP_034600.2 | 4 | 455 | |||
GenBank mRNA | AK004938.1 | - | 4 | 432 | ||
AK028181.1 | - | 4 | 433 | |||
AK043260.1 | - | 3 | 231 | |||
AK050234.1 | - | 4 | 433 | |||
AK150784.1 | - | 4 | 500 | |||
AK161041.1 | - | 3 | 225 | |||
AK161084.1 | - | 4 | 349 | |||
AK169740.1 | - | 4 | 361 | |||
BC020024.1 | - | 4 | 405 | 3597151 | ||
BC107687.1 | - | 4 | 390 | |||
BY130345.1 | - | 3 | 292 | |||
CN670198.1 | - | 3 | 296 | |||
X99642.1 | - | 2 | 98 |