Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hp1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122896.2 | NP_001116368.1 | 12 | 2736 | ||
NM_001122897.2 | NP_001116369.1 | 13 | 2973 | |||
NM_001285478.1 | NP_001272407.1 | 13 | 2993 | |||
NM_001285479.1 | NP_001272408.1 | 13 | 2931 | |||
NM_001285480.1 | NP_001272409.1 | 12 | 2697 | |||
NM_001285481.1 | NP_001272410.1 | 13 | 2895 | |||
NM_010470.3 | NP_034600.2 | 13 | 2934 | |||
GenBank mRNA | AK004938.1 | - | 13 | 2911 | ||
AK043260.1 | - | 12 | 2710 | |||
AK050234.1 | - | 13 | 2913 | |||
AK075725.1 | - | 12 | 2674 | |||
AK150784.1 | - | 13 | 2976 | |||
AK161041.1 | - | 12 | 2704 | |||
AK166161.1 | - | 13 | 2881 | |||
AK169740.1 | - | 13 | 2840 | |||
BC004053.1 | - | 9 | 2209 | |||
BC020024.1 | - | 13 | 2885 | 3597151 | ||
CF423826.1 | - | 1 | 98 |