Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd17b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010475.1 | NP_034605.1 | 6 | 1036 | ||
GenBank mRNA | AK019406.1 | - | 3 | 554 | ||
AK143346.1 | - | 6 | 985 | |||
BC125657.1 | - | 6 | 1035 | 40130172 | ||
BC125659.1 | - | 6 | 1035 | 40130174 | ||
X89627.1 | - | 6 | 1036 |