Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elavl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010487.2 | NP_034617.1 | 2 | 589 | ||
GenBank mRNA | AK090281.1 | - | 2 | 589 | ||
BC052097.1 | - | 2 | 576 | 5695543 | ||
U29148.1 | - | 2 | 568 |