Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ica1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252266.1 | NP_001239195.1 | 3 | 307 | ||
NM_010492.3 | NP_034622.3 | 4 | 720 | |||
GenBank mRNA | AB303567.1 | - | 3 | 231 | ||
AK005965.1 | - | 4 | 425 | |||
AK016989.1 | - | 4 | 423 | |||
AK192988.1 | - | 4 | 337 | |||
AK206264.1 | - | 2 | 100 | |||
BC002030.1 | - | 4 | 354 | 3487814 | ||
CF914747.1 | - | 4 | 423 | |||
DV659030.1 | - | 2 | 92 | |||
U26459.1 | - | 4 | 343 | |||
U26460.1 | - | 3 | 249 | |||
U26461.1 | - | 3 | 248 | |||
U34595.1 | - | 2 | 216 |