Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010494.1 | NP_034624.1 | 3 | 779 | ||
GenBank mRNA | AK007801.1 | - | 3 | 714 | ||
BC039128.1 | - | 3 | 711 | 3962648 | ||
BC039970.1 | - | 3 | 731 | 4487248 | ||
X65493.1 | - | 3 | 779 |