Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111320.1 | NP_001104790.1 | 3 | 566 | ||
NM_010497.3 | NP_034627.3 | 3 | 544 | |||
GenBank mRNA | AF020039.1 | - | 2 | 394 | ||
AK087063.1 | - | 3 | 446 | |||
AK149019.1 | - | 3 | 475 | |||
AK151212.1 | - | 3 | 469 | |||
AK159173.1 | - | 3 | 475 | |||
AK160896.1 | - | 3 | 476 | |||
AK167158.1 | - | 3 | 516 | |||
AK167583.1 | - | 3 | 480 | |||
AK168832.1 | - | 3 | 518 | |||
BC088986.1 | - | 3 | 516 | 6808731 | ||
CF950262.1 | - | 3 | 505 | |||
DQ403075.1 | - | 2 | 294 |