Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111320.1 | NP_001104790.1 | 6 | 971 | ||
NM_010497.3 | NP_034627.3 | 6 | 949 | |||
GenBank mRNA | AF020039.1 | - | 5 | 799 | ||
AK087063.1 | - | 6 | 851 | |||
AK149019.1 | - | 6 | 880 | |||
AK151212.1 | - | 6 | 874 | |||
AK159173.1 | - | 6 | 880 | |||
AK160896.1 | - | 6 | 881 | |||
AK167158.1 | - | 6 | 921 | |||
AK167583.1 | - | 6 | 885 | |||
AK168832.1 | - | 6 | 923 | |||
BC088986.1 | - | 6 | 921 | 6808731 | ||
DQ403075.1 | - | 5 | 699 |