Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Idh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111320.1 | NP_001104790.1 | 8 | 1229 | ||
NM_010497.3 | NP_034627.3 | 8 | 1207 | |||
GenBank mRNA | AF020039.1 | - | 7 | 1057 | ||
AK087063.1 | - | 8 | 1109 | |||
AK149019.1 | - | 8 | 1138 | |||
AK151212.1 | - | 8 | 1132 | |||
AK159173.1 | - | 8 | 1138 | |||
AK160896.1 | - | 8 | 1139 | |||
AK167158.1 | - | 8 | 1179 | |||
AK167583.1 | - | 8 | 1143 | |||
AK168832.1 | - | 8 | 1180 | |||
AK216686.1 | - | 2 | 63 | |||
BC088986.1 | - | 8 | 1179 | 6808731 | ||
DQ403075.1 | - | 7 | 957 |