Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281748.1 | NP_001268677.1 | 21 | 3149 | ||
NM_001281749.1 | NP_001268678.1 | 20 | 3025 | |||
NM_025164.4 | NP_079440.3 | 20 | 3205 | |||
XM_005271481.2 | XP_005271538.1 | 21 | 3387 | |||
XM_005271482.4 | XP_005271539.2 | 20 | 3243 | |||
XM_005271484.3 | XP_005271541.2 | 20 | 3063 | |||
XM_005271485.3 | XP_005271542.1 | 8 | 1725 | |||
XM_011542721.2 | XP_011541023.1 | 21 | 3349 | |||
XM_011542722.1 | XP_011541024.1 | 21 | 3207 | |||
XM_011542723.2 | XP_011541025.1 | 21 | 3230 | |||
XM_011542724.2 | XP_011541026.1 | 20 | 3084 | |||
XM_011542725.2 | XP_011541027.1 | 19 | 2967 | |||
XM_017017424.1 | XP_016872913.1 | 19 | 2919 | |||
XM_017017425.1 | XP_016872914.1 | 21 | 3425 | |||
XM_017017426.1 | XP_016872915.1 | 19 | 3299 | |||
XM_017017427.1 | XP_016872916.1 | 16 | 2702 | |||
GenBank mRNA | AB209327.1 | - | 13 | 2215 | ||
AK022302.1 | - | 2 | 760 | |||
AK122838.1 | - | 6 | 2467 | |||
AY598338.1 | - | 2 | 757 | |||
BC035583.2 | - | 3 | 1035 | |||
BC063887.1 | - | 6 | 1218 | |||
BC086858.1 | - | 2 | 760 | |||
BC128510.1 | - | 20 | 2887 | 40081639 | ||
BC128511.1 | - | 21 | 3005 | 40081640 |